Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec® Thulium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
VeriSpec® Thulium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
  RCRV010631100N
 :  
undefined
VeriSpec® Thulium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
  RCRV010631100N
 :  RV010631-100N
 :  

 

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010631100N
  • Element:
    Thulium
  • Supplier no.:
    RV010631-100N

 

 

Certified reference material (CRM) for ICP-MS

 : Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility