VeriSpec® Sulfur Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
Catalog # RCRV010617100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:10 µg/ml
- Matrix:H₂O
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010617100N
- Element:Sulfur
- Supplier no.:RV010617-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility