Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO?/HF
VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO?/HF
Catalog # RCRV010592100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO?/HF
Catalog # RCRV010592100N
Supplier:  Ricca Chemical
Supplier Number:  RV010592-100N
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃/HF
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010592100N
  • Element:
    Hafnium
  • Supplier no.:
    RV010592-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility