Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec® Tellurium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
VeriSpec® Tellurium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Catalog # RCRV010559100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Tellurium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Catalog # RCRV010559100N
Supplier:  Ricca Chemical
Supplier Number:  RV010559-100N
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010559100N
  • Element:
    Tellurium
  • Supplier no.:
    RV010559-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility