VeriSpec® Silicon Standard for ICP/MS 100 ppm in H₂O, RICCA Chemical Company
Catalog # RCRV010553100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:H₂O
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010553100N
- Element:Silicon
- Supplier no.:RV010553-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility