Order Entry
Puerto Rico
ContactUsLinkComponent
VeriSpec™ Manganese Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
VeriSpec™ Manganese Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
  RCRV010534100N
 :  
undefined
VeriSpec™ Manganese Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
  RCRV010534100N
 :  RV010534-100N
 :  

 

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RCRV010534100N
  • Element:
    Manganese
  • Supplier no.:
    RV010534-100N

 

 

Certified reference material (CRM) for ICP-MS

 : Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility