VeriSpec™ Gallium Standard for ICP/MS 100 ppm in 2% HNO₃
Catalog # RCRV010521100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:2% HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. No.:RCRV010521100N
- Element:Gallium
- Supplier no.:RV010521-100N
Specifications
About this item
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility