Thermometer Holder, Chemglass
10034-322
Small Business Enterprise
- Description:Thermometer Holder, Polypropylene
- Cat. No.:10034-322
- Supplier no.:CG-3511-01
The molded polypropylene thermometer holder clips onto beakers and other vessels and can hold a thermometer from OD 5 to 11 mm.