Order Entry
Puerto Rico
ContactUsLinkComponent
Palladium(II) nitrate 5000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Irritant
Health
Toxic
Palladium(II) nitrate 5000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Catalog # RCRV010138-50N
Supplier:  Ricca Chemical
CAS Number:  
undefined
Irritant
Health
Toxic
Palladium(II) nitrate 5000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Catalog # RCRV010138-50N
Supplier:  Ricca Chemical
Supplier Number:  RV010138-50N
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 x 50 mL
  • Cat. No.:
    RCRV010138-50N
  • Supplier no.:
    RV010138-50N

Specifications

About this item

 :
 :230.43
 :GHS07
 :GHS08
 :GHS06
CAS Number:10102-05-3
Formula:N₂O₆Pd
MDL Number:MFCD00011169