LaboPlast® Thread Adapter Set, Bürkle
10018-855
- Description:LaboPlast® Thread Adapter Set
- Cat. No.:10018-855
- Supplier no.:0600-0050
The LaboPlast® thread adapter set has a standard outer thread of ¾".
- Standard thread size
- Contains four adapters
- Hexagonal adapter key
Adapter set contains Hunersdoff, Kautex, cylindrical R, and conical R thread adapters and a hexagonal key.