Orders LinkContactUsLinkComponent
AdvanBlock™-PF Blocking Solution, Advansta
AdvanBlock™-PF Blocking Solution, Advansta
AdvanBlock™ protein free block solution, 1x200 mL
Catalog # 103254-748
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  R-03023-D20
AdvanBlock™-PF Blocking Solution, Advansta
Catalog # 103254-748
Supplier:  ADVANSTA INC MS
Marketsource
CAS Number:  
AdvanBlock™-PF Blocking Solution, Advansta
AdvanBlock™ protein free block solution, 1x200 mL
CAS Number:  
Price
$243.46
Log in for availability
Each (200ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Non-protein blocking solution for fluorescent and chemiluminescent Western blotting
  • Lower background for certain antibodies that require a protein-free solution
  • Optimized to stabilize fluorescence of WesternBright MCF fluorescent secondary antibodies
  • Compatible with fluorescent and chemiluminescent detection
AdvanBlock-PF is a protein-free blocking buffer, optimized for reducing non-specific background in fluorescent and chemiluminescent Western blots. It acts quickly for shorter membrane incubation times, efficiently prevents non-specific binding, and stabilizes the fluorescence of secondary antibody conjugates. AdvanBlock-PF can reduce background when used with primary antibodies that have a high degree of cross-reactivity with protein blockers such as BSA, casein or milk protein. With low-quality primary antibodies that may require protein-based blocking agents, BSA or non-fat dry milk can be dissolved directly in the AdvanBlock-PF solution used to dilute the antibody.
Provided as a 5X concentrate.
Specifications
Specifications
  • Description:
    AdvanBlock™ protein free block solution
  • Size:
    1x200 mL