Orders LinkContactUsLinkComponent
Avant™ Buffer Pouches, Advansta
Avant™ Buffer Pouches, Advansta
Marketsource
Supplier:   ADVANSTA INC MS
Avant™ Buffer Pouches, Advansta
Supplier:  ADVANSTA INC MS
Marketsource
Certificates
Avant™ Buffer Pouches, Advansta
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Convenient, fast, and pure buffer preparation.
  • Molecular grade buffers of highest purity
  • Conveniently pre-measured powder in sealed pouches
  • Quickly dissolves for immediate use
  • Standardized to guarantee reproducible results for your gels and blots
Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-measured molecular biology-grade chemicals in 500 ml of deionized water to obtain a ready-to-use 1x buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.
One pouch dissolved in 500ml purified water provides 1X working buffer.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 103254-886
    Avant™ Buffer Pouches - PBS
    Avant™ Buffer Pouches - PBS
    1xbox of 20 pouch
    Log in for availability
    Log in for availability
    Each
    Each - $76.15
    $76.15
    Each
       $76.15
    Supplier Number: R-01038-020
    Specifications:
    Marketsource
    Marketsource
  • 103254-938
    Avant™ Buffer Pouches - TBS
    Avant™ Buffer Pouches - TBS
    1xbox of 20 pouch
    Log in for availability
    Log in for availability
    Each
    Each - $76.15
    $76.15
    Each
       $76.15
    Supplier Number: R-01039-020
    Specifications:
    Marketsource
    Marketsource
  • 103254-940
    Avant™ Buffer Pouches - Tris-Glycine SDS PAGE buffer
    Avant™ Buffer Pouches - Tris-Glycine SDS PAGE buffer
    1xbox of 20 pouch
    Log in for availability
    Log in for availability
    Each
    Each - $76.15
    $76.15
    Each
       $76.15
    Supplier Number: R-01036-020
    Specifications:
    Marketsource
    Marketsource