ContactUsLinkComponent
Forced Convection Temperature Oven, Yamato
Forced Convection Temperature Oven, Yamato
Supplier:  Yamato Scientific
Certificates
undefined
Forced Convection Temperature Oven, Yamato
Supplier:  Yamato Scientific
Specifications
  • Catalog No:
  • 75846-524
  • 75846-398
  • 75846-522
  • 75846-528
  • 75846-400
  • 75846-526
  • 75846-402
  • 75846-340
  • Insulation:
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Glass Wool
  • Max. Load Per Shelf:
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • ~15 kg
  • Sensor:
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • K Thermocouple
  • Timer:
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes
  • 1 Minute to 99 Hours, 59 Minutes and 100 Hours to 999 Hours, 50 Minutes