Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD4
- Antigen Symbol:CD4
- Clonality:Monoclonal
- Clone:EP96
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Turkey,Chicken
- Size:0.5 mg
- Form:Liquid
- Gene ID:395362
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Chicken splenocytes
- Cat. No.:100245-462
- Supplier no.:8255-02
Specifications
About this item
CD4 is a type I transmembrane glycoprotein expressed on approximately 70% of thymocytes, 10% of spleen cells, and 45% of peripheral blood lymphocytes but less than 1% of bursal cells.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: EP96
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken, Turkey