Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M
- Antigen Symbol:IgM
- Clonality:Monoclonal
- Clone:UHB
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Liquid
- Gene ID:3507
- Storage Buffer:Supplied in PBS
- Storage Temperature:–20 °C
- Concentration:0.5 mg/mL
- Shipping Temperature:Dry Ice
- Tested Applications:Stim
- Cat. No.:101442-896
- Supplier no.:9022-14
Specifications
About this item
Low Endotoxin/Azide Free Anti-Human IgM antibody for use in ELISA, flow cytometry, western blotting, and stimulation assays.
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: UHB
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human