Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CD45.2
- Antigen Symbol:CD45.2
- Clonality:Monoclonal
- Clone:104
- Conjugation:RPE (R-Phycoerythrin)/Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.1 mg
- Form:Liquid
- Gene ID:19264
- Antigen Synonyms:SJL|T200|LCA|Ly-5.2
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:B10.S mouse thymocytes and splenocytes
- Cat. No.:101442-786
- Supplier no.:1800-17
The CD45 antigen is an essential regulator of leukocyte activation and development and is now known to be a protein tyrosine phosphatase, e.g., CD45 is able to dephosporylate the Src family of protein kinases. This activity is known to be required for signal transduction induced by T and B cell antigen receptor engagement, cytokine signaling, and Fc receptor stimulation. The functional state of CD45 also has a pronounced effect on lymphocyte development. CD45 exists in multiple forms, such as the restricted variants (CD45R; see companion products 1660, 1665, 1675, 1685 and 1820) and as allelic forms CD45.1 (Ly-5.1) and CD45.2 (Ly-5.2). The specificity of antibodies to CD45.1 and CD45.2 has been exploited in studies where resolution of donor and recipient cells is essential, such as in gene therapy or bone marrow transplantation. The monoclonal antibody 104 recognizes CD45 (leukocyte common antigen) on all leukocytes of mouse strains expressing the CD45.2 allotype (e.g., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C57BR, C57L, C58, DBA/1, DBA/2, NZB, SWR, 129).
Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 104
Conjugation: RPE (R-Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse