Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD79b molecule
- Antigen Symbol:CD79b
- Clonality:Monoclonal
- Clone:CB3-1
- Conjugation:Spectral Red® RPE (R-Phycoerythrin)/Cy5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:974
- Antigen Synonyms:B29|Igβ
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Ramos B cells
- Cat. No.:100246-386
- Supplier no.:9710-13
Specifications
About this item
CD79 is a disulfide-linked heterodimer composed of an α chain (CD79a) and a β chain (CD79b) that associates non-covalently with membrane immunoglobulin to form the B cell receptor (BCR) complex. Its expression is restricted to B lymphocytes first appearing on the surface at the pro-B cell stage and remaining through all stages of B cell differentiation prior to plasma cells. Crosslinking of the BCR leads to B cell activation. The monoclonal antibody CB3-1 reacts with the β chain of CD79.
Type: Primary
Antigen: CD79b
Clonality: Monoclonal
Clone: CB3-1
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human