Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin M
- Antigen Symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:1.0 mg
- Cross Adsorption:Cross-adsorbed against rat IgG for minimal reactivity
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:1.0 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Rat IgM
- Tested Applications:FLISA
- Purification:Pooled antisera from goats hyperimmunized with rat IgM; purified by affinity chromatography on pooled rat IgM covalently linked to agarose; reacts with the heavy chain of rat IgM; cross-adsorbed against rat IgG for minimal reactivity
- Cat. No.:100244-440
- Supplier no.:3020-02
Specifications
About this item
Goat Anti-Rat IgM-FITC antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat