Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:H-2Db
- Antigen Symbol:H-2Db
- Clonality:Monoclonal
- Clone:27-11-13S
- Conjugation:Biotin
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:14964
- Antigen Synonyms:MHC Class I
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:BDF1 mouse splenocytes
- Cat. No.:100244-118
- Supplier no.:1913-08
Specifications
About this item
The monoclonal antibody 27-11-13S recognizes with the α3 domain of H-2Db class I MHC antigen. The antibody cross-reacts with the α3 domain of H-2Dd, H-2Dq, H-2Ld and H-2Lq but not H-2Kb. Reactivity with haplotypes k, f, p, r, and s has not been observed.
Type: Primary
Antigen: H-2Db
Clonality: Monoclonal
Clone: 27-11-13S
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse