CD11b, the integrin αM subunit, combines with CD18, the integrin β2 subunit, to form the integrin Mac-1, also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1/CD54. Mac-1 is expressed at varying levels on granulocytes, macrophages, dendritic cells, NK cells, and B-1 cells in the peritoneal and pleural cavities. Mac-1 expression is rapidly up-regulated on neutrophils after activation and in the same time period that CD62L is shed from the cell surface. In addition to its role in binding C3bi on opsonized targets and mediation of the subsequent ingestion process, Mac-1 is important as an adhesion molecule in the transendothelial migration of monocytes and neutrophils. Cross-reaction of M1/70 to human CD11b has been observed on monocytes, polymorphonuclear leukocytes, and NK cells.
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human, Rhesus, Baboon, Rabbit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 100243-236
- 100243-234
- Antigen Symbol:
- CD11b
- CD11b
- Antigen Name:
- CD11b
- CD11b
- Antigen Synonyms:
- C3biR|Mo1|Integrin αM|CR3|Mac-1|Mac-1α
- C3biR|Mo1|Integrin αM|CR3|Mac-1|Mac-1α
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- RPE (R-Phycoerythrin)
- RPE (R-Phycoerythrin)
- Clone:
- M1/70
- M1/70
- Reactivity:
- Baboon, Rabbit, Human, Mouse
- Baboon, Rabbit, Human, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16409
- 16409
- Isotype:
- IgG2b
- IgG2b
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- C57BL/10 mouse splenic T cells and concanavalin A-activated C57BL/10 splenocytes
- C57BL/10 mouse splenic T cells and concanavalin A-activated C57BL/10 splenocytes
- Form:
- Liquid
- Liquid
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- Supplied in PBS/NaN3 and a stabilizing agent
- Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:
- 2-8°C
- 2-8°C
- Shipping Temperature:
- Ambient Temperature
- Ambient Temperature