To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD3epsilon
- Antigen Symbol:CD3epsilon
- Clonality:Monoclonal
- Clone:500-A2
- Conjugation:Spectral Red® RPE (R-Phycoerythrin)/Cy5®
- Flow Cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:12501
- Antigen Synonyms:CD3 epsilon
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Immune complexes isolated from C6VL-BS thymoma cells
- Cat. No.:100243-086
- Supplier no.:1531-13
Specifications
About this item
CD3ε, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chains ranging in size from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). CD3ε is expressed on all T cells of all mouse strains. CD3 plays a major role in signaling during antigen recognition, leading to T-cell activation. 500-A2 recognizes an epitope on the 25 kDa ε chain of the CD3/TCR complex of mouse strains tested.
Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 500-A2
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse