Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-G-CSF Rat Monoclonal Antibody [clone: BVD11-37G10]
Anti-G-CSF Rat Monoclonal Antibody [clone: BVD11-37G10]
Catalog # 100242-358
Anti-G-CSF Rat Monoclonal Antibody [clone: BVD11-37G10]
Catalog # 100242-358
Supplier Number:  10128-01
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    G-CSF
  • Clonality:
    Monoclonal
  • Clone:
    BVD11-37G10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    0.5 mg
  • Form:
    Liquid
  • Gene ID:
    1440
  • Antigen Synonyms:
    macrophage-granulocyte inducer-2|G-CSA|C17orf33|Granulocyte-colony stimulating factor|pluripoietin-β|CSF3|granulocytic neutrophil colony stimulating activity|CSF3OS|MGI-2|CSF-β
  • Storage Buffer:
    Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ice Packs
  • Immunogen:
    E. coli-expressed human G-CSF
  • Tested Applications:
    Drug Delivery
  • Cat. No.:
    100242-358
  • Supplier no.:
    10128-01

Specifications

About this item

Purified Anti-Human G-CSF antibody for use in immunohistochemistry / immunocytochemistry, ELISA, and drug delivery assays.

Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: BVD11-37G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human