To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD45RA molecule
- Antigen Symbol:CD45RA
- Clonality:Monoclonal
- Clone:F8-11-13
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:0.1 mg
- Form:Liquid
- Gene ID:5788
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Purified lymphocytes from human lymph nodes
- Tested Applications:Purification
- Cat. No.:100246-278
- Supplier no.:9630-01
Specifications
About this item
CD45RA is a 220 kDa exon A-dependent isoform of CD45. It is expressed on 40-50% of CD4+ peripheral T cells, 50% of peripheral CD8+ T cells, and a portion of B lymphocytes and monocytes.
Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: F8-11-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human