Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD15 molecule
- Antigen Symbol:CD15
- Clonality:Monoclonal
- Clone:28
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:2526
- Antigen Synonyms:3-fucosyl-N-acetyllactosamine|Lewis X|SSEA-1|3-FAL
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Monocytes separated from other peripheral leukocytes on fibronectin plates
- Cat. No.:100246-182
- Supplier no.:9565-02
Specifications
About this item
CD15 is a carbohydrate antigen found on the cell surface of neutrophils and monocytes, but not on basophils, platelets, lymphocytes, or erythrocytes. CD15 may function in adhesion as its ligand is L-selectin (CD62L).
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: 28
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human