Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD3 molecule
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:UCHT1
- Conjugation:RPE (R-Phycoerythrin)/Cy5.5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:916
- Antigen Synonyms:Leu-4|T3|TCRE|T3E|CD3ε
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Human peripheral blood lymphocytes from a patient with Sézary's syndrome and infant thymocytes
- Cat. No.:100246-044
- Supplier no.:9515-16
Specifications
About this item
CD3, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chains ranging from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). CD3 is expressed on 70-80% of normal peripheral blood lymphocytes and on 10-20% of thymocytes. It plays a major role in signaling during antigen recognition, leading to T cell activation. The monoclonal antibody UCHT1 reacts with the 20 kDa ε chain of the CD3/TCR complex.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: RPE (R-Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human