Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UCHT1]
 
Anti-CD3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: UCHT1]

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • 103260-172
  • 100246-034
  • Antigen Symbol:
  • CD3
  • CD3
  • Antigen Name:
  • CD3 molecule
  • CD3 molecule
  • Antigen Synonyms:
  • Leu-4|T3|TCRE|T3E|CD3ε
  • Leu-4|T3|TCRE|T3E|CD3ε
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • RPE (R-Phycoerythrin)
  • RPE (R-Phycoerythrin)
  • Clone:
  • UCHT1
  • UCHT1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Human peripheral blood lymphocytes from a patient with Sézary's syndrome and infant thymocytes
  • Human peripheral blood lymphocytes from a patient with Sézary's syndrome and infant thymocytes
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • Lot specific
  • Lot specific
  • Storage Buffer:
  • Supplied in PBS/NaN3 and a stabilizing agent
  • Supplied in PBS/NaN3 and a stabilizing agent
  • Storage Temperature:
  • 2-8°C
  • 2-8°C
  • Shipping Temperature:
  • Ambient Temperature
  • Ambient Temperature