Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD19 molecule
- Antigen Symbol:CD19
- Clonality:Monoclonal
- Clone:SJ25-C1
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:0.5 mg
- Form:Liquid
- Gene ID:930
- Antigen Synonyms:Leu-12|B-lymphocyte antigen|B4
- Storage Buffer:Supplied in PBS
- Storage Temperature:–20 °C
- Concentration:0.5 mg/mL
- Shipping Temperature:Dry Ice
- Immunogen:NALM-1 and NALM-16 leukemia cell line
- Tested Applications:Block
- Cat. No.:100245-902
- Supplier no.:9340-14
Specifications
About this item
CD19 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily. It is expressed at all stages of B cell differentiation except terminally differentiated plasma cells. It is also present on the cell surface of follicular dendritic cells. CD19 associates with CD21, CD81, Leu 13 and/or MHC Class II molecules to form a signal transduction complex on the B cell surface. This signaling complex modulates the activation threshold for the B cell antigen receptor (BCR).
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25-C1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human