Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD11a molecule
- Antigen Symbol:CD11a
- Clonality:Monoclonal
- Clone:38
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:3683
- Antigen Synonyms:integrin αL|ITGAL|LFA-1α
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Fibronectin purified monocytes
- Cat. No.:100245-878
- Supplier no.:9320-02
Specifications
About this item
CD11a, also known as leukocyte function-associated antigen-1 (LFA-1α), represents the 180 kDa integrin αL subunit which combines with the CD18 β2 integrin subunit to form the αLβ2 integrin heterodimer. It is expressed on lymphocytes, granulocytes, monocytes, and macrophages and is upregulated on activated T cells. CD11a mediates adhesion of lymphocytes to vascular endothelium and is involved in costimulation.
Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: 38
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human