Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G3 Hinge
- Antigen Symbol:IgG3 Hinge
- Clonality:Monoclonal
- Clone:HP6050
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Liquid
- Gene ID:3502
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Human IgG3 myeloma protein
- Tested Applications:ELISPOT, Multiplex
- Cat. No.:100245-828
- Supplier no.:9210-08
Specifications
About this item
Biotin Anti-Human IgG3 Hinge antibody for use in ELISA, flow cytometry, immunohistochemistry, western blotting, immunoprecipitation, ELISpot, and multiplex assays.
Type: Secondary
Antigen: IgG3 Hinge
Clonality: Monoclonal
Clone: HP6050
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human