Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin A1
- Antigen Symbol:IgA1
- Clonality:Monoclonal
- Clone:B3506B4
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:1.0 mL
- Form:Liquid
- Gene ID:3493
- Storage Buffer:Supplied in a stock solution of 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as a preservative
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Fc fragment of human IgA1 myeloma protein
- Tested Applications:ELISPOT
- Cat. No.:100245-758
- Supplier no.:9130-04
Specifications
About this item
AP Anti-Human IgA1 antibody for use in ELISA, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.
Type: Secondary
Antigen: IgA1
Clonality: Monoclonal
Clone: B3506B4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human