Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G F(c)
- Antigen Symbol:IgG F(c)
- Clonality:Monoclonal
- Clone:H2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Mangabey,African green monkey,Cynomolgus monkey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Tested Applications:FLISA
- Cat. No.:100245-694
- Supplier no.:9042-02
Specifications
About this item
FITC Anti-Human IgG Fc antibody for use in flow cytometry and western blotting assays.
Type: Secondary
Antigen: IgG F(c)
Clonality: Monoclonal
Clone: H2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rhesus, Cynomolgus, Chimpanzee, African Green Monkey, Mangabey, Pigtail