Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin D
- Antigen Symbol:IgD
- Clonality:Monoclonal
- Clone:IADB6
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:1.0 mL
- Form:Liquid
- Gene ID:3495
- Storage Buffer:Supplied in a stock solution of 50% glycerol/50% PBS, pH 7.4; No preservative added
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Human IgD myeloma protein
- Cat. No.:100245-676
- Supplier no.:9030-05
Specifications
About this item
HRP Anti-Human IgD antibody for use in ELISA and immunohistochemistry assays.
Type: Secondary
Antigen: IgD
Clonality: Monoclonal
Clone: IADB6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human