Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Bu-1b
- Antigen Symbol:Bu-1b
- Clonality:Monoclonal
- Clone:5-11G2
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken
- Size:0.1 mg
- Form:Liquid
- Gene ID:396098
- Antigen Synonyms:ChB6.2
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Bursa cells from two- to five-week-old CHA (Bu-1b) strain chickens
- Cat. No.:100245-580
- Supplier no.:8370-09
Specifications
About this item
Chicken Bu-1b, a product of the Bu-1b allele, is expressed on the cell surface as a disulfide-linked homodimer. In 10-week-old H.B15 and H.B2 chickens, Bu-1 is found on 85-90% of bursal cells, 2-8% of thymocytes, 15-27% of spleen cells, and 2-18% of peripheral blood cells. It is also expressed on subsets of macrophages and monocytes but not on granulocytes, erythrocytes, or thrombocytes. In chickens heterozygous for the Bu-1 alleles (Bu-1a/b), Bu-1a does not exhibit allelic exclusion. The monoclonal antibody 5-11G2 reacts with lymphoid cells from CHA and H.B14Ab strains but does not react with cells from H.B14Aa
Type: Primary
Antigen: Bu-1b
Clonality: Monoclonal
Clone: 5-11G2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken