Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:beta-2-Microglobulin
- Clonality:Monoclonal
- Clone:F21-21
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Turkey,Chicken
- Western Blot:Yes
- Size:0.5 mg
- Form:Liquid
- Gene ID:414830
- Antigen Synonyms:B-F|MHC Class I
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Affinity purified chicken MHC-I molecules
- Tested Applications:Purification
- Cat. No.:100245-550
- Supplier no.:8355-01
Specifications
About this item
Like their mammalian counterparts, avian MHC Class I molecules, also known as B-F antigens, consist of a highly polymorphic α-chain noncovalently bound to the invariant β2-microglobulin subunit. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesized antigenic peptides to CD8+ T lymphocytes which are usually cytotoxic T cells. The monoclonal antibody F21-21 also reacts with turkey β2-microglobulin.
Type: Primary
Antigen: β2-Microglobulin
Clonality: Monoclonal
Clone: F21-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Turkey