Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD8
- Antigen Symbol:CD8
- Clonality:Monoclonal
- Clone:fCD8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Lion,Tsushima leopard cat,Cat
- Size:0.5 mg
- Form:Liquid
- Gene ID:493948/493799
- Antigen Synonyms:fCD8
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Feline thymocytes
- Tested Applications:Sep
- Cat. No.:100245-392
- Supplier no.:8120-01
Specifications
About this item
Feline CD8, a member of the immunoglobulin superfamily of cell surface receptors, is a type II transmembrane glycoprotein that is expressed as a heterodimer on the suppressor/cytotoxic subpopulation of peripheral T lymphocytes. It is present on approximately 76% of thymocytes, 14% of splenocytes, 1% of bone marrow cells, and 15% of peripheral blood lymphocytes. CD8 functions as a co-receptor with MHC Class I-restricted T cell receptors in antigen recognition.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: fCD8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Feline, Lion, Tsushima Leopard Cat