Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rhesus monkey
- Size:1.0 mg
- Epitope:Heavy and Light Chain
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:1.0 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Rhesus IgG
- Tested Applications:FLISA
- Purification:Pooled antisera from goats hyperimmunized with rhesus IgG; purified by affinity chromatography on rhesus IgG covalently linked to agarose; reacts with the heavy and light chains of rhesus IgG
- Cat. No.:100245-206
- Supplier no.:6200-03
Goat Anti-Rhesus IgG(H+L)-TRITC antibody for use in immunocytochemistry assays.
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rhesus