Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:1.0 mg
- Epitope:Heavy and Light Chain
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:1.0 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Mouse IgG
- Tested Applications:FLISA
- Purification:Pooled antisera from rabbits hyperimmunized with mouse IgG; purified by affinity chromatography on mouse IgG covalently linked to agarose; reacts with the heavy and light chains of mouse IgG and the light chains of mouse IgM and IgA
- Cat. No.:100245-164
- Supplier no.:6170-02
Specifications
About this item
Rabbit Anti-Mouse IgG(H+L)-FITC antibody for use in immunohistochemistry and western blotting assays.
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse