Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G2a
- Antigen Symbol:IgG2a
- Clonality:Monoclonal
- Clone:2A8F4
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:0.1 mg
- Form:Liquid
- Gene ID:367586
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Rat IgG2a hybridoma
- Tested Applications:FLISA
- Cat. No.:100244-602
- Supplier no.:3065-09
PE Anti-Rat IgG2a antibody for use in flow cytometry assays.
Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: 2A8F4
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat