Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD49e Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5H10.27]
Anti-CD49e Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5H10.27]
Catalog # 100243-574
Anti-CD49e Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5H10.27]
Catalog # 100243-574
Supplier Number:  1695-09

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD49e
  • Antigen Symbol:
    CD49e
  • Clonality:
    Monoclonal
  • Clone:
    5H10.27
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    16402
  • Antigen Synonyms:
    VLA-5α|Integrin α5
  • Storage Buffer:
    Supplied in PBS/NaN3 and a stabilizing agent
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.1 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    C57BL/6 x A/J F1 mouse mast cell line MC/9
  • Cat. No.:
    100243-574
  • Supplier no.:
    1695-09

Specifications

About this item

The monoclonal antibody 5H10.27 reacts with the α5 chain (VLA-5) of the CD49e/CD29 heterodimeric fibronectin receptor. It is expressed on thymocytes, activated T cells, mast cells, and a variety of mouse cell lines. Soluble 5H10.27 antibody inhibits VLA-5-mediated functions in vitro. Immobilized 5H10.27 has also been demonstrated to costimulate the proliferative response of CD8+ T cells to plate-bound anti-CD3 monoclonal antibody.

Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: 5H10.27
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse