Order Entry
United States
ContactUsLinkComponent
 
Anti-CD3 epsilon Hamster Monoclonal Antibody [clone: 500-A2]
  100243-076
Anti-CD3 epsilon Hamster Monoclonal Antibody [clone: 500-A2]
  100243-076
 :  1531-01
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD3epsilon
  • Antigen Symbol:
    CD3epsilon
  • Clonality:
    Monoclonal
  • Clone:
    500-A2
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Hamster
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Size:
    0.5 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    12501
  • Antigen Synonyms:
    CD3 epsilon
  • Storage Buffer:
    Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Immune complexes isolated from C6VL-BS thymoma cells
  • Tested Applications:
    Activ
  • Cat. No.:
    100243-076
  • Supplier no.:
    1531-01

 

 

CD3ε, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chains ranging in size from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). CD3ε is expressed on all T cells of all mouse strains. CD3 plays a major role in signaling during antigen recognition, leading to T-cell activation. 500-A2 recognizes an epitope on the 25 kDa ε chain of the CD3/TCR complex of mouse strains tested.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 500-A2
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse