Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphotyrosine
- Antigen Symbol:Phosphotyrosine
- Clonality:Monoclonal
- Clone:PY20
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:1.0 mL
- Form:Liquid
- Antigen Synonyms:pTyr20
- Storage Buffer:Supplied in a stock solution of 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as a preservative
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Phosphotyrosine conjugated to carrier proteins
- Cat. No.:100242-990
- Supplier no.:1400-04
Specifications
About this item
Protein tyrosine residues are phosphorylated as a result of intracellular protein kinase activation (e.g., via growth factors) during normal growth and development and in oncogenesis. The most abundant population of target proteins for tyrosine phosphorylation is cell surface glycoproteins. Antibodies to phosphotyrosine enable the detection, isolation, and characterization of proteins containing phosphotyrosine. The monoclonal antibody PY20 prevents internalization of activated receptors (e.g., EGFR) when microinjected into cells. The affinity of PY20 for phosphotyrosine is approximately 10-6 to 10-7 M. PY20 binding to phosphorylated tyrosines can be inhibited by free phosphotyrosine and phenylphosphate but not by phosphoserine, phosphothreonine, or free phosphate.
Type: Primary
Antigen: Phosphotyrosine
Clonality: Monoclonal
Clone: PY20
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: