Order Entry
Puerto Rico
ContactUsLinkComponent
Saline 0.9%, Hardy Diagnostics
Saline 0.9%, Hardy Diagnostics
Catalog # 75860-368
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Saline 0.9%, Hardy Diagnostics
Catalog # 75860-368
Supplier:  Hardy Diagnostics
Supplier Number:  U264
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Saline 0.9%, HDx
  • Cat. No.:
    75860-368
  • Format:
    Bottle
  • Volume:
    500 ml
  • Supplier no.:
    U264

Specifications

About this item

Saline 0.9% for sterility testing filter rinsing.

  • Sterility testing filter rinse
Ordering information: Minimum Order of 28 Packs.