ContactUsLinkComponent
Urea R Broth, Hardy Diagnostics
Urea R Broth, Hardy Diagnostics
Catalog # 75860-366
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Urea R Broth, Hardy Diagnostics
Catalog # 75860-366
Supplier:  Hardy Diagnostics
Supplier Number:  Y141
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Urea R Broth, HDx
  • Cat. No.:
    75860-366
  • Tube size:
    13×100 mm
  • Supplier no.:
    Y141
Specifications
About this item
Urea R Broth.
For the rapid detection of urease activity by microorganisms. "R" is for "Rapid".
Ordering information: Minimum Order of 5 Packs.