Orders LinkContactUsLinkComponent
ELISABright, Advansta
Marketsource
Supplier:   ADVANSTA INC MS
ELISABright, Advansta
Supplier:  ADVANSTA INC MS
Marketsource
Certificates
ELISABright, Advansta
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Superior chemiluminescent substrate for ELISA applications.
  • Highest signal-to-noise ratio for superior performance
  • Broad dynamic range for enhanced detection and precision
  • Immediate light generation for fast detection and results
  • Cost effective with reduced reagent consumption
ELISABright is an enhanced and highly sensitive chemiluminescent HRP substrate optimized for chemiluminescent ELISA applications. This luminol-based substrate demonstrates a wide linear dynamic range (Figure 1) and a superior signal to noise ratio (Figure 2) for excellent quantitative ability. ELISABright allows for enhanced detection of low abundance proteins and reduced consumption of antibodies and other reagents.
 : Includes ELISABright luminol and ELISABright peroxide reagents.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 103254-876
    ELISABright Substrate
    ELISABright Substrate
    100 mL
    Log in for availability
    Log in for availability
    Each
    Each - $340.38
    $340.38
    Each
       $340.38
    Supplier Number: K-16025-D10
    Specifications:
    Marketsource
    Marketsource
  • 103254-920
    ELISABright Substrate
    ELISABright Substrate
    250 mL
    Log in for availability
    Log in for availability
    Each
    Each - $579.23
    $579.23
    Each
       $579.23
    Supplier Number: K-16025-D25
    Specifications:
    Marketsource
    Marketsource