SHOWA Atlas 330 Natural Rubber Coated Work Glove with Reinforced Thumb Crotch, Showa
Supplier: Showa
Certificates
About this item
When a job requires added protection from abrasive materials, this glove offers additonal protection with a reinforced thumb crotch.
- Reinforced thumb crotch for longer wear and protection
- Machine washable for longer wear
- Ergonmic design for comfort and dexterity
Designed with a reinfoced thumb crotch for long lasting comfort and a natural rubber coating, SHOWA 300 gloves provide top notch abrasion resistance and grip, no matter how tough the task.
Ordering information: Order by Master case of 144 pair.
Packaging: Packaged 12 pair per poly bag, 12 bags per Master case.
Specifications
- Catalog No:
- 10847-730
- 10847-726
- Disposable/Reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex
- Latex
- Liner Material:
- Cotton/Polyester
- Cotton/Polyester
- Finish:
- Natural Rubber
- Natural Rubber
- Gauge:
- 10 Gauge Seamless Knitted Liner
- 10 Gauge Seamless Knitted Liner