SHOWA Atlas 300B Natural Rubber Coated General-Purpose Work Glove, Showa
Supplier: Showa
Certificates
About this item
When handling extremely abrasive items and working in dirty environments, this glove provides longer wear and more protection than alternatives
- Dark color hides grime
- Rough coating provides extra grip
- Cotton/poly liner for added comfort
Featuring a black natural rubber palm coating and dark gray, 10 gauge polyester and cotton liner, SHOWA 300B gloves are the perfect fit for any dirty general-purpose job.
Ability to launder extends use.
Ordering information: Order by Master case of 144 pair.
Packaging: Packaged 12 pair per poly bag, 12 bags per Master case.
Specifications
- Catalog No:
- 89208-306
- 89208-302
- 89208-308
- 89208-304
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner Material:
- Cotton/Polyester
- Cotton/Polyester
- Cotton/Polyester
- Cotton/Polyester
- Finish:
- Natural Rubber Coating, Wrinkle Finish
- Natural Rubber Coating, Wrinkle Finish
- Natural Rubber Coating, Wrinkle Finish
- Natural Rubber Coating, Wrinkle Finish
- Gauge:
- 10 Gauge Seamless Liner
- 10 Gauge Seamless Liner
- 10 Gauge Seamless Liner
- 10 Gauge Seamless Liner