Order Entry
United States
ContactUsLinkComponent
DRG® Chlamydia trachomatis IgA ELISA, DRG International
DRG® Chlamydia trachomatis IgA ELISA, DRG International
Catalog # 75871-174
Supplier:  DRG International
CAS Number:  
undefined
DRG® Chlamydia trachomatis IgA ELISA, DRG International
Catalog # 75871-174
Supplier:  DRG International
Supplier Number:  EIA-3461R
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type (ELISA with LOV):
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Description:
    Chlamydia trachomatis IgA ELISA
  • Environmentally Preferable:
  • Sample Type:
    Serum
  • Application:
    Infectious Diseases
  • Regulatory Status:
    FDA Regulated; CE Approved
  • Cat. No.:
    75871-174
  • Supplier no.:
    EIA-3461
  • No. of tests:
    96 wells

Specifications

About this item

An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Chlamydia trachomatis in serum and plasma.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

Chlamydiae are non-motile, Gram negative and obligatory intracellular growing bacteria which form characteristic inclusions within the cytoplasm of parasitized cells

The DRG Chlamydia trachomatis IgA ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Patient samples are diluted with Sample Diluentand additionally incubated withIgG-RF-Sorbentto eliminate competitive inhibition from specific IgG. This pretreatment avoids false negative results. Microtiter wells as a solid phase are coated with inactivated Elementary Bodies Chlamydia trachomatis antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Chlamydia trachomatis-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgA antibodies are dispensed into the wells. During a second incubation this anti‑IgA conjugate binds specifically to IgA antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Chlamydia trachomatis-specific IgA antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.

Chlamydia trachomatis is the most prevalent agent of sexually transmitted diseases worldwide (400-500 million cases) and the number of infections is constantly growing. Rates in sexually active young people are commonly between 5% and 10% in Europe. In women, Chlamydia trachomatis can lead to pelvic inflammatory desease (PID), tubal infertility and ectopic pregnancy. Infection during pregnancy is associated with premature rupture of the membranes, low birth weight and miscarriage. Chlamydia trachomatis can also be transmitted from mother to baby during labor, causing eye and respiratory infections. In men, Chlamydia trachomatis can lead to acute genital inflammation (epididymitis, epididymo-orchitis) and occasionally to sexually-acquired reactive arthritis (SARA). In men and women, Chlamydia trachomatis may produce proctitis. Individuals with Chlamydia trachomatis are at increased risk of acquiring or transmitting HIV. Extraarticular infection with Chlamydia trachomatis caused an inflammatory reactive arthritis (Chlamydia-induced arthritis [CIA]). A severe problem in Chlamydia infections is the frequent asymptomatic insidiuous course which may result in the initiation of chronic diseases. In many instances primary infections are not recognized and only the sequelae caused by ascended, persisting agents are diagnosed. Infection can be indentified by Microscopy (Giemsa stain), PCR, Serology: Detection of antigens by ELISA, detection of antibodies by IF, EIA, ELISA. After primary infection, IgM, IgA, and IgG antibodies can be detected successively in serum samples. IgG antibodies are generally considered as markers for any contact with the pathogen irrespective of disease stage. IgM antibodies are characteristic for acute infection and IgA antibodies indicate ongoing progression of an infection.