Order Entry
United States
ContactUsLinkComponent
DRG® Androstenedione ELISA, DRG International, Inc.
DRG® Androstenedione ELISA, DRG International, Inc.
  75871-324
 :  
undefined
DRG® Androstenedione ELISA, DRG International, Inc.
  75871-324
 :  EIA-3265
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Assay Type (ELISA with LOV):
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Description:
    Androstenedione ELISA
  • Environmentally Preferable:
  • Sample Type:
    Serum
  • Detection Range:
    0.019 ng/ml
  • Application:
    Fertility/Pregnancy
  • Regulatory Status:
    Not FDA Regulated; CE Approved
  • Cat. No.:
    75871-324
  • Supplier no.:
    EIA-3265
  • No. of tests:
    96 wells

 

 

An enzyme immunoassay for the quantitative measurement of androstenedione in serum and plasma.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

The steroid hormone Androstenedione is one of the main androgens, besides Testosterone and Dehydroepiandrosterone

The DRG Androstenedione ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site of the Androstenedione molecule. Endogenous Androstenedione of a patient sample competes with an Androstenedione-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of Androstenedione in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of Androstenedione in the patient sample.

Testosterone, the most important biological active androgen, is derived from peripheral enzymatic conversion of Androstenedione. In males, androgens are secreted primarily by the Leydig cells of the testes, to some degree also in the adrenal cortex. In females, the androgens are secreted mainly in the adrenal glands and in the ovary. Around 10% of the androgens are derived from peripheral conversion, mainly of DHEA. Androstenedione and Testosterone show high diurnal variability. The highest levels are measured in the morning. At the age of puberty serum androstenedione levels rise, after menopause they decline again. High androstenedione levels are measured during pregnancy. In women, high levels of androstenedione (47-100% above normal) are generally found in hirsutism, mostly in combination with other androgens as testosterone and DHEA-S. Androstenedione overproduction is due to ovarian dysfunction or may be of adrenal origin. High circulating androstenedione levels are found in women with polycystic ovaries and 21-hydroxylase effect. Significant lower androstenedione levels are found in postmenopausal osteoporosis.