Order Entry
Puerto Rico
ContactUsLinkComponent
DRG® Thyroid Stimulating Hormone (TSH) ELISA, DRG International
DRG® Thyroid Stimulating Hormone (TSH) ELISA, DRG International
Catalog # 75871-272
Supplier:  DRG International
CAS Number:  
undefined
DRG® Thyroid Stimulating Hormone (TSH) ELISA, DRG International
Catalog # 75871-272
Supplier:  DRG International
Supplier Number:  EIA-1782
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    Thyroid Stimulating Hormone
  • Description:
    Thyroid Stimulating Hormone (TSH) ELISA
  • Sample Type:
    Serum
  • Application:
    Thyroid Function
  • Regulatory Status:
    FDA Regulated; CE Approved
  • Cat. No.:
    75871-272
  • Supplier no.:
    EIA-1782
  • No. of tests:
    96 wells

Specifications

About this item

An enzyme immunoassay for the quantitative determination of thyroid stimulating hormone (TSH) in serum.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

The determination of serum or plasma levels of thyroid stimulating hormone (TSH or thyrotropin) is recognized as a sensitive method in the diagnosis of primary and secondary hypothyroidism

The TSH ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay. The assay system utilizes a unique monoclonal antibody directed against a distinct antigenic determinant on the intact TSH molecule. Mouse monoclonal anti-TSH antibody is used for solid phase immobilization (microtiter wells), and goat anti-TSH antibody is in the antibody-enzyme (horseradish peroxidase) conjugate solution. The test sample is allowed to react simultaneously with the two antibodies, resulting in the TSH molecules being sandwiched between the solid phase and enzyme-linked antibodies. After a 60-minute or overnight incubation at room temperature, the solid phase is washed with water to remove unbound labeled antibodies. A solution of 3,3’,5,5’-Tetramethylbenzidine (TMB) is added and incubated for 20 minutes, resulting in the development of a blue color. The color development is stopped with the addition of 1 N HCl, and the resulting yellow color is measured spectrophotometrically at 450 nm. The concentration of TSH is directly proportional to the color intensity of the test sample.

TSH is secreted by the anterior lobe of the pituitary gland and induces the production and release of thyroxine (T4) and triiodothyronine (T3) from the thyroid gland. It is a glycoprotein with a molecular weight of approximately 28,000 daltons, consisting of two chemically different subunits, alpha and beta. Although the concentration of TSH in the blood is extremely low, it is essential for the maintenance of normal thyroid function. The release of TSH is regulated by a TSH-releasing hormone (TRH) produced by the hypothalamus. The levels of TSH and TRH are inversely related to the level of thyroid hormone. When there is a high level of thyroid hormone in the blood, less TRH is released by the hypothalamus, so less TSH is secreted by the pituitary. The opposite action will occur when there is decreased thyroid hormone in the blood. This process is known as a negative feedback mechanism and is responsible for maintaining the proper blood levels of these hormones. TSH and the pituitary glycoproteins: luteinizing hormone (LH), follicle-stimulating hormone (FSH), and human chorionic gonadotropin (hCG), have identical alpha chains. The beta chains are distinct but do contain regions with identical amino acid sequences. These regions of homology can cause considerable cross-reactivity with some polyclonal TSH antisera. The use of a monoclonal antibody in this TSH ELISA test eliminates such cross-reactivity, which could result in falsely elevated TSH values in either menopausal or pregnant females - a population whose evaluation of thyroid status is clinically significant.