MP Biomedicals' Instrument Platform is intended for use in conjunction with the Consumables Platforms and Test Kits.
- Choose between a solid or liquid instrument platform, depending on sample type
- Platforms support any combination of testing endpoints
- System requires minimal bench space
- Results in as little as 30 minutes
The Liquid Sample Instrument Platforms contain all of the instrumentation necessary to run liquid samples using MP Biomedicals' Free Fatty Acid, Malonaldehyde, Percent Fat, Total Glycerin, Alkenal and Peroixde Test Kits
The Solid Sample Instrument Platforms contain all of the instrumentation necessary to run solid samples using MP Biomedicals' Free Fatty Acid, Malonaldehyde, Percent Fat, Total Glycerin, Peroxide or Alkenal Test Kits. This Solid Sample Instrument Platform requires the use of MP Biomedicals' Solid Consumables Platform.
This Liquid Sample Instrument Platform requires the use of MP Biomedicals' Liquid Sample Consumables Platform.
30 x 27.2 x 12.5 cm (analyzer only)
- Detection Method:Bichromatic photometer
- Storage Temperature:RT
- Application:Oil/Fat Degradation, Biodiesel